4-Methoxy-6-oxo-1,6-dihydropyridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A1127723-1G A1127723-250MG
76672-668EA 281.14 USD
76672-668 76672-670
4-Methoxy-6-oxo-1,6-dihydropyridine-3-carboxylic acid 95%
4-Methoxy-6-oxo-1,6-dihydropyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₄
MW: 169.1348 g/mol
MDL Number: MFCD28667427
CAS Number: 57658-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR