8-Bromooct-1-yne 95%

Supplier: Ambeed
Warning

A1127719-100MG A1127719-250MG A1127719-1G A1127719-5G
76672-664EA 44.56 USD
76672-664 76672-666 77549-184 77570-700
8-Bromooct-1-yne 95%
8-Bromooct-1-yne

Precaución: for Research Use Only

Formula: C₈H₁₃Br
MW: 189.0928 g/mol
MDL Number: MFCD20621358
CAS Number: 81216-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR