N2,N4,N6-Trimethyl-1,3,5-triazine-2,4,6-triamine 95%

Supplier: Ambeed
Warning

A1127285-1G A1127285-250MG A1127285-5G
76672-652EA 196 USD
76672-652 76672-654 76672-656
N2,N4,N6-Trimethyl-1,3,5-triazine-2,4,6-triamine 95%
N2,N4,N6-Trimethyl-1,3,5-triazine-2,4,6-triamine

Precaución: for Research Use Only

Formula: C₆H₁₂N₆
MDL Number: MFCD01715762
CAS Number: 2827-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR