3-Carbamoylbenzene-1-sulfonyl fluoride 95%

Supplier: Ambeed
Danger

A1127048-250MG A1127048-1G
76672-644EA 80 USD
76672-644 77783-263
3-Carbamoylbenzene-1-sulfonyl fluoride 95%
3-Carbamoylbenzene-1-sulfonyl fluoride

Precaución: for Research Use Only

Formula: C₇H₆FNO₃S
MW: 203.1908 g/mol
MDL Number: MFCD24632641
CAS Number: 454-94-4
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR