Sodium 2,2-dimethylpropane-1-sulfinate 98%

Supplier: Ambeed
Warning

A1126111-100MG A1126111-1G A1126111-250MG
76672-626EA 50 USD
76672-626 76672-628 76672-630
Sodium 2,2-dimethylpropane-1-sulfinate 98%
Sodium 2,2-dimethylpropane-1-sulfinate

Precaución: for Research Use Only

Formula: C₅H₁₁NaO₂S
MDL Number: MFCD28166512
CAS Number: 51928-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR