Sodium 2H-1,3-benzodioxole-5-sulfinate 95%

Supplier: Ambeed
Warning

A1125913-100MG A1125913-1G A1125913-250MG
76672-612EA 119.89 USD
76672-612 76672-614 76672-616
Sodium 2H-1,3-benzodioxole-5-sulfinate 95%
Sodium 2H-1,3-benzodioxole-5-sulfinate

Precaución: for Research Use Only

Formula: C₇H₅NaO₄S
MW: 208.1670 g/mol
MDL Number: MFCD29958536
CAS Number: 253594-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR