Sodium 2,3-dihydro-1h-indene-5-sulfinate 95%

Supplier: Ambeed
Warning

A1125893-100MG A1125893-1G A1125893-250MG
76672-606EA 34 USD
76672-606 76672-608 76672-610
Sodium 2,3-dihydro-1h-indene-5-sulfinate 95%
Sodium 2,3-dihydro-1h-indene-5-sulfinate

Precaución: for Research Use Only

Formula: C₉H₉NaO₂S
MW: 204.2213 g/mol
MDL Number: MFCD24113302
CAS Number: 153261-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR