3-{[(4-fluorophenyl)methyl]amino}benzamide 95%

Supplier: Ambeed
Warning

A1125611-100MG A1125611-1G
76672-572EA 28 USD
76672-572 76672-574
3-{[(4-fluorophenyl)methyl]amino}benzamide 95%
3-{[(4-fluorophenyl)methyl]amino}benzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₃FN₂O
MDL Number: MFCD11159266
CAS Number: 1042534-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR