Ethyl 2-(3-chloropyridin-2-yl)-5-oxopyrazolidine-3-carboxylate 95%

Supplier: Ambeed
Warning

A1124707-100G A1124707-1G A1124707-250MG A1124707-25G A1124707-5G
76672-554EA 1308.09 USD
76672-554 76672-556 76672-558 76672-560 76672-562
Ethyl 2-(3-chloropyridin-2-yl)-5-oxopyrazolidine-3-carboxylate 95%
Ethyl 2-(3-chloropyridin-2-yl)-5-oxopyrazolidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClN₃O₃
MW: 269.6843 g/mol
MDL Number: MFCD09955582
CAS Number: 500011-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR