3-(Fluorosulfonyl)-4-methoxybenzoic acid 95%

Supplier: Ambeed
Danger

A1123926-250MG A1123926-100MG
76672-550EA 126 USD
76672-550 77783-256
3-(Fluorosulfonyl)-4-methoxybenzoic acid 95%
3-(Fluorosulfonyl)-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₅S
MDL Number: MFCD00024852
CAS Number: 199461-16-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR