3-Bromo-1-(4-fluorophenyl)-1H-pyrazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A1121914-100MG A1121914-1G
76672-528EA 51.99 USD
76672-528 76672-530
3-Bromo-1-(4-fluorophenyl)-1H-pyrazole-5-carboxylic acid 95%
3-Bromo-1-(4-fluorophenyl)-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrFN₂O₂
MW: 285.0692 g/mol
MDL Number: MFCD16683349
CAS Number: 1251249-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR