3,4-Dichlorobenzene-1-carboximidamide hydrochloride 97%

Supplier: Ambeed
Warning

A1121751-1G A1121751-5G
76672-524EA 22 USD
76672-524 76672-526
3,4-Dichlorobenzene-1-carboximidamide hydrochloride 97%
3,4-Dichlorobenzene-1-carboximidamide hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇Cl₃N₂
MDL Number: MFCD22369995
CAS Number: 50292-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR