3-Ethynyl-N-(thiophen-2-ylmethyl)aniline 95%

Supplier: Ambeed
Warning

A1121248-1G A1121248-250MG
76672-512EA 165.51 USD
76672-512 76672-514
3-Ethynyl-N-(thiophen-2-ylmethyl)aniline 95%
3-Ethynyl-N-(thiophen-2-ylmethyl)aniline

Precaución: for Research Use Only

Formula: C₁₃H₁₁NS
MW: 213.2981 g/mol
MDL Number: MFCD11138203
CAS Number: 1019586-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR