5-tert-Butyl-1,2,4-triazin-3-amine 95%

Supplier: Ambeed
Warning

A1117902-100MG A1117902-1G A1117902-250MG
76672-504EA 27 USD
76672-504 76672-506 76672-508
5-tert-Butyl-1,2,4-triazin-3-amine 95%
5-tert-Butyl-1,2,4-triazin-3-amine

Precaución: for Research Use Only

Formula: C₇H₁₂N₄
MW: 152.1970 g/mol
MDL Number: MFCD12131301
CAS Number: 937-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR