Quinoline-5-sulfonamide 95%

Supplier: Ambeed
Warning

A1116836-100MG A1116836-1G A1116836-250MG
76672-490EA 63.66 USD
76672-490 76672-492 76672-494
Quinoline-5-sulfonamide 95%
Quinoline-5-sulfonamide

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂S
MW: 208.2370 g/mol
MDL Number: MFCD02752662
CAS Number: 415913-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR