5,6-Dimethoxyindole 98%
Supplier: Ambeed
|
Warning
|
A111612-100MG
A111612-10G
A111612-1G
A111612-250MG
A111612-25G
A111612-5G
76672-476EA
21.22
USD
76672-476
76672-478
76672-480
76672-482
76672-484
76672-486
5,6-Dimethoxyindole 98%
5,6-Dimethoxyindole
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO₂ MW: 177.2 g/mol |
MDL Number:
MFCD00005675 CAS Number: 14430-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...