2,4-Dichloro-5-fluoroquinoline 95%

Supplier: Ambeed
Warning

A1116079-250MG A1116079-1G
76672-474EA 187.78 USD
76672-474 77619-864
2,4-Dichloro-5-fluoroquinoline 95%
2,4-Dichloro-5-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₄Cl₂FN
MW: 216.0392 g/mol
MDL Number: MFCD29089457
CAS Number: 2092047-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR