Ethyl 6-bromo-4-fluoro-1H-indole-2-carboxylate 95%

Supplier: Ambeed
Warning

A1114739-100MG A1114739-1G A1114739-250MG A1114739-5G
76672-456EA 61.54 USD
76672-456 76672-458 76672-460 76672-462
Ethyl 6-bromo-4-fluoro-1H-indole-2-carboxylate 95%
Ethyl 6-bromo-4-fluoro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉BrFNO₂
MW: 286.0971 g/mol
MDL Number: MFCD12412804
CAS Number: 396075-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR