Sodium 2-(1,3-benzothiazol-2-yl)acetate 95%

Supplier: Ambeed
Warning

A1114574-250MG A1114574-1G
76672-452EA 89.12 USD
76672-452 77619-854
Sodium 2-(1,3-benzothiazol-2-yl)acetate 95%
Sodium 2-(1,3-benzothiazol-2-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₆NNaO₂S
MW: 215.2042 g/mol
MDL Number: MFCD15209626
CAS Number: 796883-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR