1-Chloropentan-2-one 95%

Supplier: Ambeed
Danger

A1114254-1G A1114254-5G
76672-446EA 153 USD
76672-446 76672-448
1-Chloropentan-2-one 95%
1-Chloropentan-2-one

Precaución: for Research Use Only

Formula: C₅H₉ClO
MDL Number: MFCD16167605
CAS Number: 19265-24-8
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR