N-(4-(4-Methoxypiperidin-1-yl)benzyl)ethanamine 95%

Supplier: Ambeed
Warning

A1113278-100MG A1113278-1G A1113278-250MG
76672-432EA 46 USD
76672-432 76672-434 76672-436
N-(4-(4-Methoxypiperidin-1-yl)benzyl)ethanamine 95%
N-(4-(4-Methoxypiperidin-1-yl)benzyl)ethanamine

Precaución: for Research Use Only

Formula: C₁₅H₂₄N₂O
MW: 248.3639 g/mol
MDL Number: MFCD11978628
CAS Number: 1096865-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR