2-Chloro-6-(2,2,3,3-tetrafluoropropoxy)pyrazine 95%

Supplier: Ambeed
Warning

A1113038-1G A1113038-250MG
76672-428EA 279.02 USD
76672-428 76672-430
2-Chloro-6-(2,2,3,3-tetrafluoropropoxy)pyrazine 95%
2-Chloro-6-(2,2,3,3-tetrafluoropropoxy)pyrazine

Precaución: for Research Use Only

Formula: C₇H₅ClF₄N₂O
MW: 244.5700 g/mol
MDL Number: MFCD11197521
CAS Number: 1039858-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR