6-Chloro-3-phenyl-1-benzofuran-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A1112798-100MG A1112798-1G A1112798-250MG
76672-422EA 51.99 USD
76672-422 76672-424 76672-426
6-Chloro-3-phenyl-1-benzofuran-2-carboxylic acid 95%
6-Chloro-3-phenyl-1-benzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₉ClO₃
MW: 272.6832 g/mol
MDL Number: MFCD13196283
CAS Number: 1208402-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR