2,4-Diiodo-6-nitroaniline 97%

Supplier: Ambeed
Warning

A1112240-100MG A1112240-1G A1112240-250MG
76672-416EA 19 USD
76672-416 76672-418 76672-420
2,4-Diiodo-6-nitroaniline 97%
2,4-Diiodo-6-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄I₂N₂O₂
MW: 389.92 g/mol
MDL Number: MFCD03931845
CAS Number: 116529-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR