2-(3,4-Dichlorophenyl)acetamide 95%

Supplier: Ambeed
Warning

A1110644-100MG A1110644-1G A1110644-250MG
76672-390EA 27 USD
76672-390 76672-392 76672-394
2-(3,4-Dichlorophenyl)acetamide 95%
2-(3,4-Dichlorophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO
MDL Number: MFCD11043647
CAS Number: 868697-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR