2-Chloro-n-(2-hydroxyethyl)-n-phenylacetamide 95%

Supplier: Ambeed
Danger

A1110395-250MG A1110395-5G
76672-386EA 19.1 USD
76672-386 76672-388
2-Chloro-n-(2-hydroxyethyl)-n-phenylacetamide 95%
2-Chloro-n-(2-hydroxyethyl)-n-phenylacetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO₂
MW: 213.6608 g/mol
MDL Number: MFCD11987075
CAS Number: 28025-61-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR