2-Amino-6-chlorobenzene-1-thiol hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A1110389-1G
A1110389-250MG
76672-382EA
217
USD
76672-382
76672-384
2-Amino-6-chlorobenzene-1-thiol hydrochloride 95%
2-Amino-6-chlorobenzene-1-thiol hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₇Cl₂NS |
MDL Number:
MFCD12197228 CAS Number: 385376-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...