3-[(2H-1,3-Benzodioxol-5-yloxy)methyl]aniline 95%

Supplier: Ambeed
Warning

A1109848-1G A1109848-250MG
76672-372EA 181 USD
76672-372 76672-374
3-[(2H-1,3-Benzodioxol-5-yloxy)methyl]aniline 95%
3-[(2H-1,3-Benzodioxol-5-yloxy)methyl]aniline

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₃
MDL Number: MFCD11131653
CAS Number: 1019395-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR