2,1,3-Benzoselenadiazol-4-amine 95%

Supplier: Ambeed
Danger

A1108808-1G A1108808-5G A1108808-100MG
76672-358EA 171 USD
76672-358 76672-360 77783-214
2,1,3-Benzoselenadiazol-4-amine 95%
2,1,3-Benzoselenadiazol-4-amine

Precaución: for Research Use Only

Formula: C₆H₅N₃Se
MDL Number: MFCD04620394
CAS Number: 767-65-7
UN: 3283
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR