2-Amino-N,N,4-trimethylpentanamide hydrochloride 95%

Supplier: Ambeed
Warning

A1108772-1G A1108772-5G
76672-354EA 255 USD
76672-354 76672-356
2-Amino-N,N,4-trimethylpentanamide hydrochloride 95%
2-Amino-N,N,4-trimethylpentanamide hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₉ClN₂O
MDL Number: MFCD11505609
CAS Number: 1214130-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR