2-(6-Nitro-1H-indazol-1-yl)acetic acid 95%

Supplier: Ambeed
Warning

A1108615-100MG A1108615-1G A1108615-250MG
76672-348EA 28.65 USD
76672-348 76672-350 76672-352
2-(6-Nitro-1H-indazol-1-yl)acetic acid 95%
2-(6-Nitro-1H-indazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₇N₃O₄
MW: 221.1696 g/mol
MDL Number: MFCD09805059
CAS Number: 857801-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR