5-(Chlorosulfonyl)-2,4-dimethylbenzoic acid 95%

Supplier: Ambeed
Danger

A1108244-1G A1108244-25G A1108244-5G
76672-332EA 259.93 USD
76672-332 76672-334 76672-336
5-(Chlorosulfonyl)-2,4-dimethylbenzoic acid 95%
5-(Chlorosulfonyl)-2,4-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₄S
MW: 248.6834 g/mol
MDL Number: MFCD09049987
CAS Number: 313346-23-5
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR