tert-Butyl 4-(4-(aminomethyl)benzyl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A1108153-1G A1108153-100MG A1108153-250MG
76672-330EA 356.47 USD
76672-330 77619-840 77619-842
tert-Butyl 4-(4-(aminomethyl)benzyl)piperazine-1-carboxylate 95%
tert-Butyl 4-(4-(aminomethyl)benzyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₇N₃O₂
MW: 305.4152 g/mol
MDL Number: MFCD30751495
CAS Number: 1415109-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR