1-((tert-Butoxycarbonyl)amino)-2-methylcyclobutanecarboxylic acid 98%

Supplier: Ambeed
Warning

A1108103-100MG A1108103-250MG A1108103-25MG
76672-320EA 292.81 USD
76672-320 76672-322 76672-324
1-((tert-Butoxycarbonyl)amino)-2-methylcyclobutanecarboxylic acid 98%
1-((tert-Butoxycarbonyl)amino)-2-methylcyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.2729 g/mol
MDL Number: MFCD24535850
CAS Number: 1860337-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR