4-(2-Bromoacetyl)benzene-1-sulfonamide 95%

Supplier: Ambeed
Danger

A1107478-250MG A1107478-5G
76672-310EA 127.31 USD
76672-310 76672-312
4-(2-Bromoacetyl)benzene-1-sulfonamide 95%
4-(2-Bromoacetyl)benzene-1-sulfonamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃S
MW: 278.1230 g/mol
MDL Number: MFCD08689427
CAS Number: 944-33-2
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR