2-(3,5-Dichloro-2-hydroxyphenyl)acetic acid 95%

Supplier: Ambeed
Warning

A1106876-1G A1106876-250MG
76672-304EA 217.49 USD
76672-304 76672-306
2-(3,5-Dichloro-2-hydroxyphenyl)acetic acid 95%
2-(3,5-Dichloro-2-hydroxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₃
MW: 221.0374 g/mol
MDL Number: MFCD22005340
CAS Number: 38692-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR