4-tert-Butyl-3-chloroaniline 95%

Supplier: Ambeed
Warning

A1106845-250MG A1106845-5G A1106845-1G
76672-302EA 18 USD
76672-302 77570-602 77783-206
4-tert-Butyl-3-chloroaniline 95%
4-tert-Butyl-3-chloroaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClN
MDL Number: MFCD09966032
CAS Number: 52756-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR