2,6-Difluoro-4-(hydroxymethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A1106791-100MG A1106791-250MG
76672-300EA 83.82 USD
76672-300 77619-836
2,6-Difluoro-4-(hydroxymethyl)benzoic acid 95%
2,6-Difluoro-4-(hydroxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
MDL Number: MFCD20657333
CAS Number: 1378805-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR