3-Cyano-5-fluorobenzene-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A1105095-1G A1105095-250MG A1105095-5G
76672-280EA 51 USD
76672-280 76672-282 76672-284
3-Cyano-5-fluorobenzene-1-sulfonyl chloride 95%
3-Cyano-5-fluorobenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃ClFNO₂S
MDL Number: MFCD18394025
CAS Number: 1261644-49-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR