1-(4-Bromo-2-nitrophenyl)-2,2,2-trifluoroethan-1-ol 95%

Supplier: Ambeed
Warning

A1104530-100MG A1104530-1G A1104530-250MG
76672-266EA 49.87 USD
76672-266 76672-268 76672-270
1-(4-Bromo-2-nitrophenyl)-2,2,2-trifluoroethan-1-ol 95%
1-(4-Bromo-2-nitrophenyl)-2,2,2-trifluoroethan-1-ol

Precaución: for Research Use Only

Formula: C₈H₅BrF₃NO₃
MW: 300.0294 g/mol
MDL Number: MFCD30727609
CAS Number: 2092313-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR