5-Bromo-2-(Ethylsulfonyl)pyridine 95%

Supplier: Ambeed
Warning

A1103817-1G A1103817-250MG A1103817-5G
76672-260EA 160.2 USD
76672-260 76672-262 76672-264
5-Bromo-2-(Ethylsulfonyl)pyridine 95%
5-Bromo-2-(Ethylsulfonyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂S
MW: 250.1129 g/mol
MDL Number: MFCD22418065
CAS Number: 223556-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR