1-Bromo-4-(2,2-dibromoethenyl)-benzene 95%

Supplier: Ambeed
Warning

A1103557-1G A1103557-250MG
76672-256EA 110 USD
76672-256 76672-258
1-Bromo-4-(2,2-dibromoethenyl)-benzene 95%
1-Bromo-4-(2,2-dibromoethenyl)-benzene

Precaución: for Research Use Only

Formula: C₈H₅Br₃
MDL Number: MFCD00273344
CAS Number: 136350-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR