1,1-Dimethoxy-N-methylpropan-2-amine 95%
Supplier: Ambeed
|
Danger
|
A1103446-100MG
A1103446-1G
A1103446-250MG
A1103446-5G
76672-242EA
74.27
USD
76672-242
76672-244
76672-246
76672-248
1,1-Dimethoxy-N-methylpropan-2-amine 95%
1,1-Dimethoxy-N-methylpropan-2-amine
Precaución: for Research Use Only
Formula:
C₆H₁₅NO₂ MW: 133.1888 g/mol |
MDL Number:
MFCD19218004 CAS Number: 65765-12-0 UN: 2924 ADR: 3,8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...