1,3-Bis(3,5-dichlorophenyl)urea 98+%

Supplier: Ambeed
Warning

A1103292-1MG A1103292-25MG A1103292-50MG
76672-236EA 11 USD
76672-236 76672-238 76672-240
1,3-Bis(3,5-dichlorophenyl)urea 98+%
1,3-Bis(3,5-dichlorophenyl)urea
Formula: C₁₃H₈Cl₄N₂O
MDL Number: MFCD01566238
CAS Number: 73439-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR