5-Cyclopropyl-1-(2-methylphenyl)-1H-pyrazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A1097723-100MG A1097723-1G A1097723-250MG
76672-218EA 54 USD
76672-218 76672-220 76672-222
5-Cyclopropyl-1-(2-methylphenyl)-1H-pyrazole-4-carboxylic acid 95%
5-Cyclopropyl-1-(2-methylphenyl)-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O₂
MDL Number: MFCD09971624
CAS Number: 241800-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR