4-Fluoro-N-(furan-2-ylmethyl)-2-methylaniline 95%

Supplier: Ambeed
Warning

A1096232-1G A1096232-250MG
76672-202EA 128 USD
76672-202 77783-188
4-Fluoro-N-(furan-2-ylmethyl)-2-methylaniline 95%
4-Fluoro-N-(furan-2-ylmethyl)-2-methylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₂FNO
MDL Number: MFCD01135302
CAS Number: 418785-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR