3-Fluoro-N-(furan-2-ylmethyl)aniline 95%

Supplier: Ambeed
Warning

A1095452-1G A1095452-250MG
76672-196EA 144.29 USD
76672-196 76672-198
3-Fluoro-N-(furan-2-ylmethyl)aniline 95%
3-Fluoro-N-(furan-2-ylmethyl)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₀FNO
MW: 191.2016 g/mol
MDL Number: MFCD01308205
CAS Number: 416862-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR