Spiro[cyclopropane-1,2'-[2h]indol]-3'(1'H)-one 95%

Supplier: Ambeed

A1094950-100MG A1094950-250MG
76672-188EA 227 USD
76672-188 76672-190
Spiro[cyclopropane-1,2'-[2h]indol]-3'(1'H)-one 95%
Spiro[cyclopropane-1,2'-[2h]indol]-3'(1'H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD01683971
CAS Number: 64053-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR