2H,3H-Thieno[3,4-b][1,4]dioxine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A1094516-1G A1094516-250MG A1094516-5G
76672-182EA 35 USD
76672-182 76672-184 76672-186
2H,3H-Thieno[3,4-b][1,4]dioxine-5-carbonitrile 95%
2H,3H-Thieno[3,4-b][1,4]dioxine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₅NO₂S
MDL Number: MFCD08060540
CAS Number: 859851-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR