5-Methylisoquinoline-1-carbonitrile 95%

Supplier: Ambeed
Warning

A1093968-100MG A1093968-250MG
76672-172EA 15 USD
76672-172 76672-174
5-Methylisoquinoline-1-carbonitrile 95%
5-Methylisoquinoline-1-carbonitrile

Precaución: for Research Use Only

Formula: C₁₁H₈N₂
MDL Number: MFCD26666968
CAS Number: 1367744-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR